Learn More
Aviva Systems Biology Corp PPP3CA Antibody - N-terminal region (ARP51337_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP51337_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPP3CA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PPP3CA. Alternate names: CALN, CALNA, CALNA1, CCN1, CNA1, PPP2BApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60551