Learn More
PPP3CA Rabbit anti-Human, Unconjugated, Clone: ABOG-16, Abnova™
Rabbit Monoclonal Antibody.
Manufacturer:  Abnova Corporation MAB19684
Catalog No. 89-336-208
Add to cart
Description
Description
PPP3CA Monoclonal antibody specifically detects PPP3CA in Human samples. It is validated for ICC, WB-Ce, IF, Flow Cyt.
Specifications
Specifications
PPP3CA
ABOG-16
protein phosphatase 3 (formerly 2B), catalytic subunit, alpha isoform
Liquid
Rabbit
IgG
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing.
Monoclonal
Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
PPP3CA
CALN, CALNA, CALNA1, CCN1, CNA1, PPP2B
A synthetic peptide corresponding to human PPP3CA
100 μg
Primary
5530