Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP3CB Mouse anti-Human, Clone: OTI2E4, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527212

Catalog No. PIMA527212


Add to cart

Description

Description

PPP3CB is a calcium-dependent, calmodulin-stimulated protein phosphatase. This subunit may have a role in the calmodulin activation of calcineurin.
Specifications

Specifications

PPP3CB
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P16298
RP11-345K20.1, CALNA2, CALNB, CNA2, PP2Bbeta, CAM-PRP catalytic subunit, calcineurin A beta, calcineurin A2, calmodulin-dependent calcineurin A subunit beta isoform, protein phosphatase 2B, catalytic subunit, beta isoform, protein phosphatase 3 (formerly 2B), catalytic subunit, beta isoform, protein phosphatase from PCR fragment H32, serine/threonine-protein phosphatase 2B catalytic subunit beta isoform
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5532
Western Blot
OTI2E4
Unconjugated
PPP3CB
Liquid
PPP3CB
Human recombit protein fragment corresponding to amino acids 23-96 of PPP3CB produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit