Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPT1 Mouse anti-Human, Clone: 1117CT11.2.1.4, Abnova™

Mouse monoclonal antibody raised against full length recombinant human PPT1.

Manufacturer:  Abnova Corporation MAB12348

Catalog No. 89-934-798


Add to cart

Description

Description

The protein encoded by this gene is a small glycoprotein involved in the catabolism of lipid-modified proteins during lysosomal degradation. The encoded enzyme removes thioester-linked fatty acyl groups such as palmitate from cysteine residues. Defects in this gene are a cause of infantile neuronal ceroid lipofuscinosis 1 (CLN1, or INCL) and neuronal ceroid lipofuscinosis 4 (CLN4). Two transcript variants encoding different isoforms have been found for this gene

Specifications

Specifications

PPT1
1117CT11.2.1.4
Mouse monoclonal antibody raised against full length recombinant human PPT1.
In PBS (0.09% sodium azide)
CLN1/INCL/PPT
Mouse
IgG1
400 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Flow Cytometry, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Flow Cytometry (1:25) Immunohistochemistry (1:25) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Liquid
PPT1
Recombinant protein corresponding to full length human PPT1.
Protein G purification
RUO
Primary
5538
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit