Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPWD1 Mouse anti-Human, Clone: OTI1B2, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525280

Catalog No. PIMA525280


Add to cart

Description

Description

Putative peptidylprolyl isomerase (PPIase). PPIases accelerate the folding of proteins. It catalyzes the cis-trans isomerization of proline imidic peptide bonds in oligopeptides. May be involved in pre-mRNA splicing.
Specifications

Specifications

PPWD1
Monoclonal
0.6 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q96BP3
peptidylprolyl isomerase domain and WD repeat-containing protein 1, spliceosome-associated cyclophilin
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23398
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI1B2
Unconjugated
PPWD1
Liquid
PPWD1
Full length human recombit protein of PPWD1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit