Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPY Mouse anti-Human, Clone: GT327, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527773

Catalog No. PIMA527773


Add to cart

Description

Description

Positive Control: human PPY-transfected 293T cells Predicted Reactivity: Rhesus Monkey (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

PPY encodes a member of the neuropeptide Y (NPY) family of peptides. The encoded 95 aa preproprotein is synthesized in the pancreatic islets of Langerhans and proteolytically processed to generate two peptide products. These products include the active pancreatic hormone of 36 aa and an icosapeptide of unknown function. This hormone acts as a regulator of pancreatic and gastrointestinal functions and may be important in the regulation of food intake. Plasma level of this hormone has been shown to be reduced in conditions associated with increased food intake and elevated in anorexia nervosa. In addition, infusion of this hormone in obese rodents has shown to decrease weight gain. Alternative splicing results in multiple transcript variants, at least one of which encodes an isoform that is proteolytically processed.
Specifications

Specifications

PPY
Monoclonal
1 mg/ml
PBS with no preservative
P01298
PPY
Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human Pancreatic Polypeptide. The exact sequence is proprietary.
Protein G
RUO
Antibody
Monoclonal
Human
Western Blot
GT327
Unconjugated
PPY
Liquid
Mouse
IgG2a
100 μl
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
5539
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit