Learn More
Tocris Bioscience™ PQ 69 Available on GSA/VA Contract for Federal Government customers only.
Potent and selective A1 antagonist/inverse agonist
$248.50 - $1046.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
535110 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5351/10
Each for $248.50
Add to cart
535150 View Documents Tocris Bioscience™
5351/50
Each for $1,046.00
Add to cart
Description
Description
Potent and selective A1 receptor inverse agonist. Exhibits high binding affinity at A1 receptor (Ki values are 0.07 and 0.96 nM for rat and human receptors respectively). Reduces basal [35S]-GTPγS binding 44.6% (IC50 = 0.19 nM). Antagonizes the effects of A1 agonist R-PIA (IC50 = 18.3 nM) and exhibits competitive antagonism on CCPA -induced tracheal contractions ex vivo. Displays 217- fold selectively over hA2A receptor and >1000-fold selectivity over hA3 receptor.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.