Learn More
Invitrogen™ PQBP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592200
Description
Immunogen sequence: MPLPVALQTR LAKRGILKHL EPEPEEEIIA EDYDDDPVDY EATRLEGLPP SWYKVFDPSC GLPYYWNADT DLVSWLSPHD PNSVVTKSAK KLRSSNADAE EKLDRSHDKS DRGHDKSDRS HEKLDRGHDK SDRGHDKSDR DRERGYDKVD RERERDRERD RDRGYDKADR EEGKERRHHR REELAPYPKS KKAVSRKDEE LDPMDPSSYS DAPRGTWSTG LPKRNEAKTG ADTTAAGPLF QQRPYPSPGA VLRANAEASR TKQQD; Positive Samples: 22Rv1, SW480, HL-60, Mouse liver, Mouse brain, Rat brain, Rat ovary, Rat liver, SKOV3; Cellular Location: Nucleus, Nucleus speckle.
PQBP1 is a nuclear polyglutamine-binding protein that contains a WW domain. Mutations in this gene are associated with X-linked mental retardation.
Specifications
PQBP1 | |
Polyclonal | |
Unconjugated | |
Pqbp1 | |
38 kDa nuclear protein containing a WW domain; JM26; MGC129091 protein; MRX2; MRX55; MRXS3; MRXS8; NPW38; polyglutamine binding protein 1; polyglutamine tract-binding protein 1; Polyglutamine-binding protein 1; PQBP1; PQBP-1; RENS1; scurfy candidate 2; Sfc2; SHS | |
Rabbit | |
Affinity chromatography | |
RUO | |
10084, 302557, 54633 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O60828, Q6PCT5, Q91VJ5 | |
Pqbp1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-265 of human PQBP1 (NP_0057011). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.