Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PQBP1 Rabbit anti-Mouse, Rat, Clone: 8D8H6, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP31607520UL
This item is not returnable.
View return policy
Description
PQBP1 Monoclonal antibody specifically detects PQBP1 in Mouse, Rat samples. It is validated for Western BlotSpecifications
PQBP1 | |
Monoclonal | |
Unconjugated | |
38 kDa nuclear protein containing a WW domain, MRX55, MRXS3, MRXS8, Npw38, NPW38mental retardation, X-linked 55, nuclear protein containing WW domain 38 kD, polyglutamine binding protein 1, Polyglutamine tract-binding protein 1, polyglutamine-binding protein 1, PQBP-1, RENS1, SHS, Sutherland-Haan X-linked mental retardation syndrome | |
Recombinant protein of human PQBP1. | |
20 μg | |
Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
10084 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
8D8H6 | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction