Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PQLC2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548512

Catalog No. PA548512


Add to cart

Description

Description

Amino acid transporter that specifically mediates the pH-dependent export of the cationic amino acids arginine, histidine and lysine from lysosomes.
Specifications

Specifications

PQLC2
Polyclonal
Unconjugated
PQLC2
Liquid
PQLC2
KLH conjugated synthetic peptide between 222-251 amino acids from the C-terminal region of human PQLC2.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q6ZP29, Q8C4N4
Lysosomal amino acid transporter 1 homolog, PQ-loop repeat-containing protein 2, PQLC2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
212555, 54896
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit