Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRAC Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $517.59
Specifications
| Antigen | PRAC |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry, ELISA, Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
Description
PRAC Polyclonal specifically detects PRAC in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| PRAC | |
| Western Blot 1:500-1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cell Biology, Tumor Suppressors | |
| PBS with 0.02% Sodium Azide | |
| 84366 | |
| Full length protein (Human) | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry, ELISA, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| RUO | |
| Human | |
| C17orf92, chromosome 17 open reading frame 92, MGC32520, PRAC1, prostate cancer susceptibility candidate, Prostate, rectum and colon expressed gene protein, small nuclear protein PRAC | |
| PRAC1 | |
| IgG | |
| Affinity Purified | |
| Reacts with human PRAC protein. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title