Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pragmin Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302675A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Catalytic activity: ATP + a [protein]-L-tyrosine = ADP + a [protein]-L-tyrosine phosphateSpecifications
Pragmin | |
Polyclonal | |
Unconjugated | |
PRAG1 | |
SGK223; Tyrosine-protein kinase SgK223; Sugen kinase 223 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
157285 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q86YV5 | |
PRAG1 | |
Between 275 and 325 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title