Learn More
PRAMEF16 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA549105
Catalog No. PA549105
Add to cart
Specifications
Specifications
PRAMEF16
Polyclonal
PBS with 0.09% sodium azide
PRAME family member 16
KLH conjugated synthetic peptide between 446-472 amino acids from the C-terminal region of human PRAMEF16.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Western Blot
Unconjugated
Liquid
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
Human