Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRAMEF2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116231
Description
PRAMEF2 Polyclonal Antibody for Western Blot
The function of this protein remains unknown.
Specifications
PRAMEF2 | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
O60811 | |
PRAMEF2 | |
KLH conjugated synthetic peptide between 388-417 amino acids from the C-terminal region of human PRAMEF2. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
PRAMEF2 | |
DJ845O24.3; PRAME family member 2; PRAMEF2 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
65122 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction