Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRAMEF2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546691
Description
Peptide sequence: VRVNWEIFTP LRAELMCTLR EFRQPKRIFI GPTPCPSCGS SPSEELELHL Sequence homology: Human: 100%.
The function of this protein remains unknown.
Specifications
PRAMEF2 | |
Polyclonal | |
Unconjugated | |
PRAMEF2 | |
DJ845O24.3; PRAME family member 2; PRAMEF2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
65122 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O60811 | |
PRAMEF2 | |
Synthetic peptide directed towards the C-terminal of human PRAMEF2 (aa 423-472). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction