Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRAMEF8 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548344

Catalog No. PA548344


Add to cart

Specifications

Specifications

PRAMEF8
Polyclonal
PBS with 0.09% sodium azide
Q5VWM4
PRAME family member 8, PRAMEF8
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
391002
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PRAMEF8
Liquid
PRAMEF8
KLH conjugated synthetic peptide between 419-447 amino acids from the C-terminal region of human PRAMEF8.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit