Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pramoxine hydrochloride, ≥98%, MilliporeSigma™ Supelco™

Pramoxine hydrochloride is a surface and local anesthetic agent , which provides relief from itching and pain for a short period of time.

$55.80 - $110.00

Specifications

Percent Purity ≥98%
Grade Analytical Standard
Linear Formula C17H27 NO3 · HCl
Physical Form Neat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-5010
View Documents
MilliporeSigma Supelco
P942210G
10 g
Each for $55.80
Add to cart
 
11-101-5009
View Documents
MilliporeSigma Supelco
P942225G
25 g
Each for $110.00
Add to cart
 
Description

Description

Pharmaceutical (small molecule)
Specifications

Specifications

≥98%
Analytical Standard
C17H27 NO3 · HCl
Neat
Safety and Handling

Safety and Handling

RTECSNumber : QD8750000

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit