Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRAP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562614
Description
Immunogen sequence: AVPAPKVPIK MQVKHWPSEQ DPEKAWGARV VEPPEKDDQL VVLFPVQKPK LLTTEEKPRG QGR Highest antigen sequence identity to the following orthologs: Mouse - 44%, Rat - 54%.
Proline rich acidic protein 1 (PRAP1) is also named as PRO1195 or UPA. It may play an important role in maintaining normal growth homeostasis in epithelial cells. Mitotic arrest deficient 1 (MAD1) is a key component in mitotic checkpoint signaling. Recent study identified PRAP1 as a novel MAD1 binding partner using yeast-two hybrid screening. PRAP1 has four isoforms with MW 16-20 kDa and 10 kDa.
Specifications
PRAP1 | |
Polyclonal | |
Unconjugated | |
PRAP1 | |
epididymis tissue protein Li 178; PRAP1; Pregnancy-specific uterine protein; PRO1195; proline rich acidic protein 1; proline-rich acidic protein 1; UNQ608/PRO1195; Upa; uterine-specific proline-rich acidic protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
118471 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96NZ9 | |
PRAP1 | |
Recombinant protein corresponding to Human PRAP1. Recombinant protein control fragment (Product #RP-104476). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction