Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRAS40 (p Thr356) Rabbit anti-Drosophila, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP260783
Description
PRAS40 Polyclonal specifically detects PRAS40 in Drosophila samples. It is validated for Western Blot.Specifications
PRAS40 (p Thr356) | |
Polyclonal | |
Western Blot 1:100-1:2000 | |
40 kDa, AKT1 substrate 1 (proline-rich), MGC2865, proline-rich AKT1 substrate 1,40 kDa proline-rich AKT substrate | |
Rabbit | |
60 kDa | |
100 μL | |
Primary | |
Drosophila | |
IgG |
Western Blot | |
Unconjugated | |
10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per ml BSA and 50% glycerol. with No Preservative | |
AKT1S1 | |
Synthetic phospho-peptide corresponding to amino acid residues surrounding Thr356 conjugated to KLH | |
Affinity Purified | |
Apoptosis, Autophagy, Cancer, Growth and Development, Lipid and Metabolism, mTOR Pathway, Neuronal Cell Markers, Neuroscience, Phospho Specific, Tumor Suppressors | |
84335 | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title