Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Praseodymium(III) acetylacetonate hydrate, +99%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
C15H21O6Pr·xH2O
MFCD08272322
C15H21O6Pr·xH2O
MFCD08272322
Assay Percent Range 99+%
Linear Formula Pr[OCH(CH3)CH2C(O)CH3]3
Formula Weight 438.24
Physical Form Powder
Infrared Spectrum Authentic
Packaging Glass bottle
Praseodymium (Pr) 99.0 min.
Color Green to White
Quantity 1g
Chemical Name or Material Praseodymium(III) acetylacetonate hydrate

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Risk of explosion if heated under confinement.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.