Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Praseodymium(III) carbonate octahydrate, 99.9% (REO), Alfa Aesar™

Manufacturer:  Alfa Aesar 1460914

Quantity 25g

 View more versions of this product

Catalog No. AA1460914


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

14948-62-0
99.9% (REO)
605.97; 461.85 (anhydrous)
Praseodymium(III) carbonate octahydrate
Crystalline
Pr2(CO3)3·8H2O
MFCD00211307

Specifications

14948-62-0
99.9% (REO)
Green crystalline
MFCD00211307
Green
Praseodymium(III) carbonate octahydrate
Crystalline
Pr2(CO3)3·8H2O
605.97; 461.85 (anhydrous)
Odorless
25g
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit