Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Praseodymium(III) chloride 99.9%, anhydrous, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
10361-79-2
Cl3Pr
MFCD00011176
acmc-20ajts,praseodymium 3+ ion trichloride,praseodymium chloride, anhydrous,praseodymium iii chloride, anhydrous, ampuled under argon trace rare earth metals basis 5g
10361-79-2
Cl3Pr
MFCD00011176
acmc-20ajts,praseodymium 3+ ion trichloride,praseodymium chloride, anhydrous,praseodymium iii chloride, anhydrous, ampuled under argon trace rare earth metals basis 5g
for ultra trace analysis
Assay Percent Range 99.90%
Linear Formula PrCl3
Formula Weight 247.27
Physical Form Powder
Grade Trace Metal
Packaging Glass bottle
Total Trace Metal Impurities 0.1% max.
Color Blue to Green
Quantity 25g
Chemical Name or Material Praseodymium(III) chloride

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.