Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Praseodymium(III) chloride hydrate, REacton™, 99.99% (REO)

Manufacturer:  Alfa Aesar™ 1123822

Quantity 100g

 View more versions of this product

Catalog No. AA1123822

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

19423-77-9
MFCD00149828
Praseodymium(III) chloride hydrate
247.27 (Anhydrous)
14,7712
Reacton™

Specifications

19423-77-9
247.27 (Anhydrous)
14,7712
115°C
Praseodymium(III) chloride hydrate
Reacton™
Wet Crystalline Aggregates
MFCD00149828
Odorless
100g
99.99%, (REO)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

RTECSNumber : TU0175000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit