Alfa Aesar™ Praseodymium(III) chloride hydrate, REacton™, 99.99% (REO)

Manufacturer: Alfa Aesar™ 1123814

Quantity 25g

 View more versions of this product

Catalog No. AA1123814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 19423-77-9
    Formula Weight 247.27 (Anhydrous)
    MDL Number MFCD00149828
    Merck Index 14,7712
    Chemical Name or Material Praseodymium(III) chloride hydrate
    Grade Reacton™

    Specifications

    Formula Weight 247.27 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P280-P264-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.
    Health Hazard 2 GHS H Statement
    H319
    Causes serious eye irritation.
    Odor Odorless
    CAS 19423-77-9
    Melting Point 115°C
    Physical Form Wet Crystalline Aggregates
    MDL Number MFCD00149828
    Merck Index 14,7712
    Quantity 25g
    Health Hazard 1 Warning
    RTECS Number TU0175000
    TSCA TSCA
    Chemical Name or Material Praseodymium(III) chloride hydrate
    Assay 99.99%, (REO)
    Grade Reacton™

    GHS H Statement
    H319
    Causes serious eye irritation.

    GHS P Statement
    P280-P264-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit