Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Praseodymium(III) chloride, ultra dry, 99.95% (REO)

Manufacturer:  Alfa Aesar™ H3781506

Quantity 5g

 View more versions of this product

Catalog No. AAH3781506


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

10361-79-2
247.27
Praseodymium(III) chloride, Ultra dry
PrCl3
MFCD00011176

Specifications

10361-79-2
PrCl3
MFCD00011176
Odorless
Hygroscopic
786°C
Praseodymium(III) chloride, Ultra dry
Powder
247.27
4.02g/mL
Ampule
1700°C
5g
99.95%, (REO)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

RTECSNumber : TU0175000

TSCA : TSCA

storageNote1 :

  • Ampouled under argon

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit