Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Praseodymium(III) hexafluoro-2,4-pentanedionate

Click to view available options
Quantity:
1g
5g
47814-20-0
C5HF6O2Pr+2
347.959
MFCD00058835
GJZQTVLINUWRRY-UHFFFAOYSA-M
praseodymium 3+ ion 1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate,1,1,1,5,5,5-hexafluoro-4-oxo-2-penten-2-olate; praseodymium 3+,1,1,1,5,5,5-hexakis fluoranyl-4-oxidanylidene-pent-2-en-2-olate; praseodymium 3+
53395444
1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate;praseodymium(3+)
C(=C(C(F)(F)F)[O-])C(=O)C(F)(F)F.[Pr+3]
47814-20-0
C5HF6O2Pr+2
347.959
MFCD00058835
GJZQTVLINUWRRY-UHFFFAOYSA-M
praseodymium 3+ ion 1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate,1,1,1,5,5,5-hexafluoro-4-oxo-2-penten-2-olate; praseodymium 3+,1,1,1,5,5,5-hexakis fluoranyl-4-oxidanylidene-pent-2-en-2-olate; praseodymium 3+
53395444
1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate;praseodymium(3+)
C(=C(C(F)(F)F)[O-])C(=O)C(F)(F)F.[Pr+3]
Formula Weight 762.06
Physical Form Powder
Quantity 1g
Chemical Name or Material Praseodymium(III) hexafluoro-2,4-pentanedionate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures