Alfa Aesar™ Praseodymium(III) isopropoxide

Manufacturer: Alfa Aesar™ 3943006

Quantity 5g

 View more versions of this product

Catalog No. AA3943006

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 19236-14-7
    Linear Formula Pr[OHC(CH3)2]3
    Formula Weight 318.12
    MDL Number MFCD00145157
    UN Number UN2925
    Chemical Name or Material Praseodymium(III) isopropoxide

    Specifications

    Formula Weight 318.12
    Health Hazard 3 GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P310
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Physical Form Powder
    Linear Formula Pr[OHC(CH3)2]3
    Melting Point 175°C (0.04mmHg) (subl.)
    Quantity 5g
    Sensitivity Moisture Sensitive
    MDL Number MFCD00145157
    CAS 19236-14-7
    UN Number UN2925
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318-H228
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable solid.
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
    Chemical Name or Material Praseodymium(III) isopropoxide

    GHS H Statement
    H314-H318-H228
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable solid.

    GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P310
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit