Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Praseodymium(III, IV) oxide, 99.9% (REO)
Click to view available options
Quantity:
10g
50g
Specifications
Solubility Information | Insoluble in water. Moderately soluble in strong mineral acids. |
Formula Weight | 1021.43 |
Physical Form | 3-12mm Sintered Lump |
Percent Purity | 99.9% |
Particle Size | 3 to 12mm |
Melting Point | 2500°C |
Quantity | 10g |
Chemical Name or Material | Praseodymium(III, IV) oxide |
Assay | (REO) |
Safety and Handling
GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 234-857-9
TSCA : Yes
Recommended Storage : Ambient temperatures