Learn More
Praseodymium(III, IV) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Description
Praseodymium(III, IV) oxide is used in pigment in glass, optic and ceramic industries. Praseodymium-doped glass, called didymium glass is used in welding, blacksmithing, and glass-blowing goggles due to its blocking property of infrared radiation. It is employed in the solid state synthesis of praseodymium molybdenum oxide, which is used as a semiconductor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 2500°C |
Color | Black |
Physical Form | Powder |
Quantity | 100 g |
Assay Percent Range | 99.99% (REO) |
Solubility Information | Insoluble in water. Moderately soluble in strong mineral acids. |
Formula Weight | 1021.43 |
Appearance | Black powder |
Chemical Name or Material | Praseodymium(III, IV) oxide |
Safety and Handling
Hazard Category | H315-H319-H335 |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 234-857-9 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.