Alfa Aesar™ Praseodymium(III, IV) oxide, 99% (REO)

Manufacturer: Alfa Aesar™ 44594A7

Quantity 5kg

 View more versions of this product

Catalog No. AA44594A7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12037-29-5
    Molecular Formula Pr6O11
    Formula Weight 1021.43
    MDL Number MFCD00011178
    Chemical Name or Material Praseodymium (III, IV) oxide

    Specifications

    Formula Weight 1021.43
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 12037-29-5
    Quantity 5kg
    Health Hazard 1 Warning
    TSCA TSCA
    Physical Form Powder
    Molecular Formula Pr6O11
    Melting Point 2500°C
    Health Hazard 2 GHS H Statement
    H319-H335
    Causes serious eye irritation.
    May cause respiratory irritation.
    MDL Number MFCD00011178
    Chemical Name or Material Praseodymium (III, IV) oxide
    Assay 99%, (REO)

    GHS H Statement
    H319-H335
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit