Learn More
Praseodymium(III) nitrate hydrate, 99.9% (REO), Thermo Scientific Chemicals
Description
Praseodymium(III) nitrate hydrate is used in fluorescent display tubes and phosphors. It is also used in the ultrasonic synthesis of phase-pure praseodymium molybdate. It plays a major role in the solvothermal process for formation of doped lanthanide oxysulfide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Green |
Physical Form | Crystalline |
Quantity | 10 g |
Assay Percent Range | 99.9% |
Solubility Information | Soluble in water,anhydrous amine,alcohol,ether,polar solvents and acetonitrile. |
IUPAC Name | praseodymium(3+) hexahydrate trinitrate |
Formula Weight | 326.92 (Anhydrous) |
Odor | Odorless |
Sensitivity | Hygroscopic |
Appearance | Green crystals |
Show More |
Safety and Handling
Hazard Category | H272-H315-H319-H335 |
Hazard Statement | In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. |
Precautionary Statement | P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
DOTInformation | DOT Class: 5.1, Packaging Group: III |
EINECSNumber | 233-796-5 |
RTECSNumber | TU1400000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.