Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Praseodymium ingot, 99.9% (REO), Alfa Aesar™

Manufacturer:  Alfa Aesar 0026709

Quantity 10g

 View more versions of this product

Catalog No. AA0026709


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7440-10-0
99.9%
Praseodymium, ingot
Solid In Various Forms
MFCD00011174

Specifications

7440-10-0
99.9%
MFCD00011174
Gray to silver
Praseodymium, ingot
Solid In Various Forms
gray to silver ingot
Odorless
10g

Safety and Handling

SDS & Safety Documents

H228-H260: Flammable solid. In contact with water releases flammable gases which may ignite spontaneously

DOTInformation : DOT Class: 4.1, Packaging Group: II

EINECSNumber : 231-120-3

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit