Alfa Aesar™ Praseodymium powder, -200 mesh, 99.9% (REO)

Manufacturer: Alfa Aesar™ 4591006

Quantity 5g

 View more versions of this product

Catalog No. AA4591006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-10-0
    MDL Number MFCD00011174
    UN Number UN3089
    Chemical Name or Material Praseodymium powder

    Specifications

    Quantity 5g
    Health Hazard 3 GHS P Statement
    P210-P231+P232-P280-P370+P378b-P402+P404-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Handle under inert gas.
    Protect from moisture.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: Special powder for metal fires.
    Store in a dry place.
    Store in a closed container.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00011174
    CAS 7440-10-0
    Storage Note 1
    • Packaged under Argon
    Sensitivity Air & Moisture Sensitive
    Color Silver
    TSCA TSCA
    UN Number UN3089
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H228-H260
    Flammable solid.
    In contact with water releases flammable gases which may ignite spontaneously.
    DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
    Mesh Size −200 Mesh
    Odor Odorless
    Physical Form Powder
    Chemical Name or Material Praseodymium powder
    Assay 99.9%, (REO)

    GHS H Statement
    H228-H260
    Flammable solid.
    In contact with water releases flammable gases which may ignite spontaneously.

    GHS P Statement
    P210-P231+P232-P280-P370+P378b-P402+P404-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Handle under inert gas.
    Protect from moisture.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: Special powder for metal fires.
    Store in a dry place.
    Store in a closed container.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under Argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit