Learn More
Sklar InstrumentsSupplier Diversity Partner Pratt Uterine Dilator, Sklar, Double End 17/19 French, 11-1/2 in., Curved, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 905219
Pratt Uterine Dilator, Sklar, Double End 17/19 French, 11-1/2 in., Curved, Stainless Steel. Sklar's Pratt Uterine Dilator is a double-ended OB/GYN instrument used for progressive dilation of the uterus. It is most frequently used during intrauterine procedures such as D&C and D&E. The size of these dilators is determined by the circumference of the tip and measured in French units, with one French unit equalling 1/3mm. Pratt dilators are available in several sizes. Each dilator has two different size tips. In comparison to other intrauterine dilators, Pratt dilators taper more gradually and are slightly lighter than Hegar dilators. They do not have a cuff to serve as a stop for depth of insertion such as featured on Hank dilators. The instrument is straight in the middle, slightly curved at each end, with a smooth, rounded tip. This product is a size 17/19 French dilator with an overall length of 11-1/2 inches.
Catalog No. 50-143-6766