Learn More
Selleck Chemical LLCSupplier Diversity Partner Pravastatin DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S571310MG
Pravastatin is a lipoprotein-lowering drug via reversibly inhibiting hydroxymethylglutaryl-CoA (HMG-CoA) reductase and the synthesis of very-low-density lipoproteins.
Catalog No. 50-194-8314