Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pravastatin Sodium, EP, 97-102%, Spectrum™

Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
81131-70-6
C23H35NaO7
446.52
VWBQYTRBTXKKOG-IYNICTALSA-M
sodium (3R,5R)-7-[(1S,2S,6S,8S,8aR)-6-hydroxy-2-methyl-8-{[(2S)-2-methylbutanoyl]oxy}-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate
[Na+].CC[C@H](C)C(=O)O[C@H]1C[C@H](O)C=C2C=C[C@H](C)[C@H](CC[C@@H](O)C[C@@H](O)CC([O-])=O)[C@@H]12
81131-70-6
C23H35NaO7
446.52
VWBQYTRBTXKKOG-IYNICTALSA-M
sodium (3R,5R)-7-[(1S,2S,6S,8S,8aR)-6-hydroxy-2-methyl-8-{[(2S)-2-methylbutanoyl]oxy}-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate
[Na+].CC[C@H](C)C(=O)O[C@H]1C[C@H](O)C=C2C=C[C@H](C)[C@H](CC[C@@H](O)C[C@@H](O)CC([O-])=O)[C@@H]12

P2882, 81131-70-6, C23H35O7Na

Spectrum™ Pravastatin Sodium, EP
Packaging Poly Pail
pH 7.2 to 9.0
Water 0.04
Quantity 5 kg
CAS Max % 1
Percent Purity 97 to 102%
Grade EP