Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRCP Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548567

Catalog No. PA548567

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Cleaves C-terminal amino acids linked to proline in peptides such as angiotensin II, III and des-Arg9-bradykinin. This cleavage occurs at acidic pH, but enzymatic activity is retained with some substrates at neutral pH.
Specifications

Specifications

PRCP
Polyclonal
PBS with 0.09% sodium azide
P42785
Lysosomal Pro-X carboxypeptidase, Angiotensinase C, Lysosomal carboxypeptidase C, Proline carboxypeptidase, Prolylcarboxypeptidase, PRCP, PRCP, PCP
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5547
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PRCP
Liquid
PRCP
KLH conjugated synthetic peptide between 114-143 amino acids from the N-terminal region of human PRCP.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit