Learn More
Aviva Systems Biology Corp PRDM1 Antibody - middle region (P100596_P050) | 100uL
Supplier:  Aviva Systems Biology Corp P100596_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRDM1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PRDM1. Alternate names: BLIMP1, MGC118922, MGC118923, MGC118924, MGC118925, PRDI-BF1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62654