Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRDM1 (BLIMP-1) Mouse anti-Human, Unconjugated, Clone: ROS195G also called 195G/G5, MilliporeSigma™

Mouse Monoclonal Antibody

$153.50 - $386.00

Specifications

Antigen PRDM1 (BLIMP-1)
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABE18141MI
View Documents
MilliporeSigma
MABE1814100UG
100 μg
Each for $386.00
Add to cart
 
MABE18142MI
View Documents
MilliporeSigma
MABE181425UG
25 μg
Each for $153.50
Add to cart
 
Description

Description

PRDM1 (BLIMP-1) Monoclonal antibody specifically detects PRDM1 (BLIMP-1) in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting.
Specifications

Specifications

PRDM1 (BLIMP-1)
Unconjugated
Mouse
PR domain zinc finger protein 1;BLIMP-1;Beta-interferon gene positive regulatory domain I-binding factor;PR domain-containing protein 1;Positive regulatory domain I-binding factor 1;PRDI-BF1;PRDI-binding factor 1
GST-tagged full-length human recombinant PR domain zinc finger protein 1.
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Purified
Human
PRDM1, BLIMP1
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit