Learn More
Aviva Systems Biology Corp PRDM10 Antibody - middle region (ARP39405_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP39405_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRDM10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PRDM10. Alternate names: KIAA1231, MGC131802, PFM7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62882