Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRDM15 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555036
Description
Immunogen sequence: KHVQLTHDKV EAQSCQLCGT KVSTRASMSR HMRRKHPEVL AVRIDDLDHL PETTTIDASS IGIVQPELTL EQEDLAEGKH GKAAKRSHKR KQKPEEEAGA PVPEDATFSE YSEKETEFTG SVGDETNSAV QS Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
The exact function of PRDM15 remains unknown.
Specifications
PRDM15 | |
Polyclonal | |
Unconjugated | |
PRDM15 | |
C21orf83; E130018M06Rik; ORF62; PFM12; PFM15; PR domain 15; PR domain containing 15; PR domain zinc finger protein 15; PR domain-containing protein 15; PR/SET domain 15; PRDM15; Zfp298; Zinc finger protein 298; ZNF298 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
63977 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P57071 | |
PRDM15 | |
Recombinant protein corresponding to Human PRDM15. Recombinant protein control fragment (Product #RP-100722). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction