Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRDM15 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP258475

Catalog No. NBP258475


Add to cart

Description

Description

PRDM15 Polyclonal antibody specifically detects PRDM15 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

PRDM15
Unconjugated
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
C21orf83, chromosome 21 open reading frame 83, PFM15, PR domain containing 15, PR domain zinc finger protein 15, PR domain-containing protein 15, Zinc finger protein 298, ZNF298
Rabbit
IgG
100 ul
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Immunocytochemistry, Immunofluorescence
Immunocytochemistry/Immunofluorescence 1-4 ug/ml
Affinity Purified
PRDM15
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:DKPMLKQAGSGVHAAGTPENSAPVESEPSQWACKVCSATFLELQLLNEHLLGHLEQAKSLPPGSQSEAAAPEKEQD
Affinity purified
RUO
Primary
63977
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only