Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRDM6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576284
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
PRDM6 contains 4 C2H2-type zinc fingers and 1 SET domain. PRDM6 may be involved in transcriptional regulation.
Specifications
PRDM6 | |
Polyclonal | |
Unconjugated | |
PRDM6 | |
Gm92; PFM3; PR domain 6; PR domain containing 6; PR domain zinc finger protein 6; PR domain-containing protein 6; PR domain-containing protein in smooth muscle; PRDM6; PR-domain zinc finger protein 6; Prism; putative histone-lysine N-methyltransferase PRDM6; RGD1559594 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
225518, 307305, 93166 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q3UZD5, Q9NQX0 | |
PRDM6 | |
Recombinant protein of human PRDM6. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction