Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRDM8 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP258446

Catalog No. NBP258446


Add to cart

Description

Description

PRDM8 Polyclonal antibody specifically detects PRDM8 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

PRDM8
Unconjugated
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
PFM5, PR domain containing 8, PR domain zinc finger protein 8, PR domain-containing protein 8, PR-domain containing protein 8
Rabbit
IgG
100 ul
Chromatin Research
Primary
56978
Human
Immunocytochemistry, Immunofluorescence, Western Blot
Western Blot 0.4 ug/ml, Immunocytochemistry/Immunofluorescence 1-4 ug/ml
Affinity Purified
PRDM8
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:RCPKRLHSADISPQDEQGGGVGTKDHGGGGGGGKDQQQQQQEAPLGPGPKFCKAGPLHHYPSPSPESSNPS
Affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only