Learn More
PRDX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 205681AP
Catalog No. 50-173-2967
Add to cart
Description
Description
The antibody is specific to PRDX1.
This gene encodes a member of the peroxiredoxin family of antioxidant enzymes, which reduce hydrogen peroxide and alkyl hydroperoxides. The encoded protein may play an antioxidant protective role in cells, and may contribute to the antiviral activity of CD8(+) T-cells. This protein may have a proliferative effect and play a role in cancer development or progression. Three transcript variants encoding the same protein have been identified for this gene.
Specifications
Specifications
PRDX1
0.48 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P35700, Q06830, Q63716
MSP23, NKEF A, NKEFA, PAG, PAGA, PAGB, peroxiredoxin 1, PRDX1, PRX1, PRXI, TDPX2, Thioredoxin peroxidase 2
Rabbit
IgG
150 μL
-20°C
Primary
117254, 18477, 5052
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
PRDX1
Liquid
Prdx1
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat