Sino Biological™ PRDX2 Recombinant Human Protein, N-His Tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding human PRDX2 (Met 1-Asn 198) fused to polyhistidine tag at N-terminus

Manufacturer: Sino Biological™ 11255H07B5

 View more versions of this product

Catalog No. 11-255-H07B5

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Molecular Weight 24kd
    Quantity 5μg
    Endotoxin Concentration <1 EU/μg
    Common Name Peroxiredoxin 2
    Gene Symbol PRDX2
    Protein Tag His Tag
    Validated Application Assay Standard
    Protein Family Other proteins
    Expression System Insect
    Product Type PRDX2 Recombinant Human Protein
    For Use With (Application) Analysis of protein structure and protein-protein interactions
    Gene ID (Entrez) 7001
    Purity ≥90% by SDS-PAGE
    Source Human
    Structural Form Full Length, Recombinant
    Format Lyophilized

    Expressed from DNA sequence encoding human PRDX2 (Met 1-Asn 198) fused to polyhistidine tag at N-terminus

    • Formulation: 50mM Tris, 100mM NaCl, 10% Glycerol, pH 8.0

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit