Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRE-084 hydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
138847-85-5
C19H27NO3
317.43
MFCD00673883
RQHKZUBCUZVZEF-UHFFFAOYSA-N
pre-084 hydrochloride, 2-morpholin-4-yl ethyl 1-phenylcyclohexane-1-carboxylate hydrochloride, pre-084, solid, 2-4-morpholinethyl 1-phenylcyclohexanecarboxylate hydrochloride, 2-4-morpholinethyl-1-phenylcyclohexanecarboxylate hydrochloride, 2-4-morpholinyl ethyl 1-phenylcyclohexanecarboxylate hydrochloride
11314197
2-(morpholin-4-yl)ethyl 1-phenylcyclohexane-1-carboxylate
O=C(OCCN1CCOCC1)C1(CCCCC1)C1=CC=CC=C1
This item is not returnable. View return policy
138847-85-5
C19H27NO3
317.43
MFCD00673883
RQHKZUBCUZVZEF-UHFFFAOYSA-N
pre-084 hydrochloride, 2-morpholin-4-yl ethyl 1-phenylcyclohexane-1-carboxylate hydrochloride, pre-084, solid, 2-4-morpholinethyl 1-phenylcyclohexanecarboxylate hydrochloride, 2-4-morpholinethyl-1-phenylcyclohexanecarboxylate hydrochloride, 2-4-morpholinyl ethyl 1-phenylcyclohexanecarboxylate hydrochloride
11314197
2-(morpholin-4-yl)ethyl 1-phenylcyclohexane-1-carboxylate
O=C(OCCN1CCOCC1)C1(CCCCC1)C1=CC=CC=C1

Highly selective σ1 agonist

High affinity, selective σ1 agonist (Ki values are 2.2 and 13091 nM for σ1 and σ2 receptors respectively). Selective over PCP receptors (IC50 > 100000 nM) and several other receptor systems. Produces antiamnesic and antitussive effects in vivo. Also exhibits protective effects in PC6.3 cells expressing mutant huntingtin by activating NF-κB signaling.
Quantity 10mg
Formula Weight 353.89
Percent Purity >99%
Chemical Name or Material PRE-084 hydrochloride

Recommended Storage : Store at Room Temperature