Learn More
Pre-BCR Rat anti-Mouse, BUV615, Clone: SL156, BD OptiBuild™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 752350
Catalog No. BDB752350
Add to cart
Description
Description
Pre-BCR Monoclonal antibody specifically detects Pre-BCR in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
Pre-BCR
Flow Cytometry
0.2 mg/mL
Ms Pre-B Cell Receptor BUV615 SL156 50 μg
Rat
IgG2a, κ
RUO
Primary
Pre-B Cell Receptor
Mouse
SL156
Brilliant Ultraviolet 615
BUV615
Purified soluble complex of recombinant µ IgH, λ5, and VpreB
1 x 50 μg
Refrigerate (2 to 8°C)
Monoclonal
Mouse