Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pre-Preserved Environmental Sample Containers

Catalog No. p-4526521
Click to view available options
Color:
Amber
Clear
Preservative:
0.2 mL 1:1 Hydrochloric Acid
0.25 mL 1:1 Hydrochloric Acid
0.5 mL 1:1 Hydrochloric Acid
10 mL Methanol, tare weighed
5 mL Methanol, tare weighed
5 mL Sodium Bisulfate

Save the time, tedium and chemical contact associated with diluting and aliquoting chemical preservatives by using our chemically-preserved sample containers.

Simplify field preservation with Thermo Scientific™ Chemically-Preserved Environmental Sample Containers and enjoy peace of mind as well as convenience. Using certified containers and chemicals reduces the risk of container and preservative contamination. Let us save you the time, tedium and chemical contact associated with diluting and aliquoting chemical preservatives to prepare environmental sample container sets. Choose from our standard offering of chemically-preserved containers, or let us custom-create container/preservative combos according to your personal analytical needs.

  • Standard Thermo Scientific Certified sample containers chemically-preserved with certified chemical preservatives for environmental sample analyses
  • Packaged in compliance with the shipping requirements of 49 CFR Part 173.4 DOT Small Quantities Classification for highway and rail transport

Ordering Information:

Custom preserved containers available by special order:

  • Choose a standard certified Thermo Scientific environmental sample container
  • Choose a chemical from our standard chemical list
  • Specify the volume and concentration of chemical to aliquot into each container

Products available only in the US

Includes:

Certified sample containers dosed with chemical preservative; certificate of analysis for container and preservative, individually barcoded containers traceable to certs, sample analysis labels unattached for optional use, hazardous material compliant packaging. MSDS available upon request.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (English) Outer 1.1 in.
Color Clear
Diameter (Metric) Outer 28 mm
Material Glass (Type 1)
Volume (Metric) 40 mL
Height (English) 3.75 in.
Level Certified
No. per Case 72
Volume (English) 1 oz.
Volume (Metric) Total 40 mL
Closure Type Septa Cap
Height (Metric) 95 mm
Thickness (Metric) 3.17 mm
Certifications/Compliance Certified to meet the EPA Performance Based Specifications for volatile organic compounds
Type Environmental Sample Container
Preservative 0.5 mL 1:1 Hydrochloric Acid
Closure Size 24-414
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.