Learn More
Athens Research And Technology IncSupplier Diversity Partner PREALBUMIN HUMAN 1MG DFS Item
Manufacturer:  Athens Research And Technology IncSupplier Diversity Partner 16161618011MG
Prealbumin, Human Plasma (Transthyretin): 1 MG; Lyophilized from 50 mM Na phosphate, pH 7.5, with 150 mM NaCl.; MW 55,000; EC= 1.35; Purity: > =95% by SDS-PAGE. This native protein is prepared and purified from human plasma.
Catalog No. 50-489-897